missing translation for 'onlineSavingsMsg'
Learn More

Rhenium(VII) oxide, 99.99% (metals basis)

Moisture Sensitive; Sublimation temperature/start: 250°C (482°F) | CAS: 1314-68-7 | O7Re2 | 484.407 g/mol

Supplier:  Thermo Scientific Chemicals 01133909

 View more versions of this product

Catalog No. AA1133909


Only null left
Explore available promotions

Description

Description

Rhenium(VII) oxide acts as a precursor to methylrhenium trioxide, organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is used as a reagent for dehydrations and rearrangements of allylic alcohols, oximes and amides, oxidative cyclizations, alkene metathesis and formation of bisperoxyacetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

360°C
297°C
White
10 g
O7Re2
rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis
NBGOSNNAAHDRLK-UHFFFAOYSA-N
trioxo(trioxorheniooxy)rhenium
123106
Odorless
Yellow powder
Rhenium(VII) oxide
1314-68-7
Crystalline
99.99% (metals basis)
MFCD00011199
Soluble in water and alcohol.
O=[Re](=O)(=O)O[Re](=O)(=O)=O
484.407
484.4
Moisture sensitive
6.103

Chemical Identifiers

1314-68-7
484.407
NBGOSNNAAHDRLK-UHFFFAOYSA-N
123106
O=[Re](=O)(=O)O[Re](=O)(=O)=O
O7Re2
MFCD00011199
rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis
trioxo(trioxorheniooxy)rhenium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 215-241-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only