CAS RN 1314-68-7
CAS RN 1314-68-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Rhenium(VII) oxide, 99.99% (metals basis)
CAS: 1314-68-7 Molecular Formula: O7Re2 Molecular Weight (g/mol): 484.407 MDL Number: MFCD00011199 InChI Key: NBGOSNNAAHDRLK-UHFFFAOYSA-N Synonym: rhenium heptoxide,rhenium vii oxide,re2o7,dirhenium heptoxide,dirhenium heptaoxide,rhenium oxide re2o7,trioxo-trioxorheniooxy rhenium,trioxo trioxorheniooxy rhenium,rhenium vii oxide-re,rhenium vii oxide trace metals basis PubChem CID: 123106 IUPAC Name: trioxo(trioxorheniooxy)rhenium SMILES: O=[Re](=O)(=O)O[Re](=O)(=O)=O