missing translation for 'onlineSavingsMsg'
Learn More

Rhenium Standard for AAS, Ready-to-Use, MilliporeSigma™ Supelco™

Catalog No. 111016886
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016886 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016886 Supplier Merck Emd Millipore Supplier No. 83672100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7697-37-2
MDL Number MFCD00011195
Synonym Rhenium nitrate solution
Type Analytical Standard
Quantity 100 mL
Grade Analytical Standard