missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGR Antibody Cocktail

Antibody Cocktail
Supplier: Invitrogen™ MA566213
Description
RGR Cocktail Antibody for IHC (P)
This gene encodes a putative retinal G-protein coupled receptor. The gene is a member of the opsin subfamily of the 7 transmembrane, G-protein coupled receptor 1 family. Like other opsins which bind retinaldehyde, it contains a conserved lysine residue in the seventh transmembrane domain. The protein acts as a photoisomerase to catalyze the conversion of all-trans-retinal to 11-cis-retinal. The reverse isomerization occurs with rhodopsin in retinal photoreceptor cells. The protein is exclusively expressed in tissue adjacent to retinal photoreceptor cells, the retinal pigment epithelium and Mueller cells. This gene may be associated with autosomal recessive and autosomal dominant retinitis pigmentosa (arRP and adRP, respectively). Alternative splicing results in multiple transcript variants encoding different isoforms.Chemical Identifiers
| RGR | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Q9Z2B3 | |
| 306307, 57811 | |
| Synthetic peptide encompassing a sequence within the Extracellular domain of human RGR. The exact sequence is proprietary. | |
| Antibody |
| HL3056+HL3058 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Cocktail | |
| Liquid | |
| IgG | |
| Mouse, Rat | |
| RGR | |
| retinal G protein coupled receptor; Rgr; RGR opsin; RGR-opsin; RP44; RPE retinal G-protein coupled receptor; RPE-retinal G protein-coupled receptor | |
| RGR | |
| Primary | |
| Protein A |
Specifications
| RGR | |
| Cocktail | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Z2B3 | |
| RGR | |
| Synthetic peptide encompassing a sequence within the Extracellular domain of human RGR. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| HL3056+HL3058 | |
| Unconjugated | |
| RGR | |
| retinal G protein coupled receptor; Rgr; RGR opsin; RGR-opsin; RP44; RPE retinal G-protein coupled receptor; RPE-retinal G protein-coupled receptor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 306307, 57811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |