missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RFX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112688
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112688 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112688 Supplier Invitrogen™ Supplier No. PA5112688
Only null left

Rabbit Polyclonal Antibody

RFX1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X2, X3, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with RFX family members X2, X3, and X5, but not with X4. This protein binds to the X-boxes of MHC class II genes and is essential for their expression. Also, it can bind to an inverted repeat that is required for expression of hepatitis B virus genes.

Specifications

Antigen RFX1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RFX1
Gene Accession No. P22670
Gene Alias AI047719; AI385641; EFC; EF-C; Enhancer factor C; MHC class II regulatory factor RFX; MHC class II regulatory factor RFX1; Regulatory factor X 1; regulatory factor X, 1 (influences HLA class II expression); regulatory factor X1; RFX; RFX1; trans-acting regulatory factor 1; transcription factor RFX1; vregulatory factor X, 1 (influences HLA class II expression)
Gene Symbols RFX1
Host Species Rabbit
Immunogen Recombinant Human MHC class II regulatory factor RFX1 protein (36-133AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5989
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less