missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RFTN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583923 Supplier Invitrogen™ Supplier No. PA583923
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: ALHPFVQPTH EREKTPLEHI FRAILIKKTD RSQKTDLHNE GYILELDCCS SLDHPTDQKL IPEFIKKIQE AASQGLKFVG VIPQYHSSV Highest antigen sequence indentity to the following orthologs: Rat - 78%, Mouse - 78%.

Raftlin, also known as RFTN1 (raftlin, lipid raft linker 1), cell migration-inducing gene 2 protein, PIB10, PIG9 or MIG2, is a 578 amino acid cell membrane protein and lipid anchor that is essential for raft cell assembly and maintenance. A member of the raftlin family, Raftlin modulates B-cell antigen receptor-mediated signaling, TCR signals, and is involved in T cell-mediated immune responses. The gene encoding Raftlin maps to human chromosome 3, which houses over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. Key tumor suppressing genes on chromosome 3 include those that encode the apoptosis mediator RASSF1, the cell migration regulator HYAL1 and the angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth Disease are a few of the numerous genetic diseases associated with chromosome 3.

Specifications

Antigen RFTN1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rftn1
Gene Accession No. Q14699
Gene Alias 2310015n21rik; AI853526; Cell migration-inducing gene 2 protein; KIAA0084; MIG2; PIB10; PIG9; proliferation-inducing protein 10; proliferation-inducing protein 9; Raftlin; raftlin lipid raft linker 1; raftlin, lipid raft linker 1; raft-linking protein; Rftn1; RGD1310777; RGD1563347
Gene Symbols Rftn1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RFTN1. Recombinant protein control fragment (Product #RP-100953).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23180
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less