missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFPL2 Monoclonal Antibody (K06336_6A2), Invitrogen™
Mouse Monoclonal Antibody
$1421.98 - $2134.69
Specifications
| Antigen | RFPL2 |
|---|---|
| Clone | K06336_6A2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: DPVGGGNLTN KRPSCAPSPQ DLSAQWKQLE DRGASSRRVD MAALFQEASS CPVCSDYLEK PMSLECGCAV CLKCINSLQK EPHGEDLLCC CSSMVSRKNK
RFPL2 Monoclonal antibody specifically detects RFPL2 in Human, Rat samples. It is validated for Western BlotSpecifications
| RFPL2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| ret finger protein like 2; ret finger protein-like 2; RFPL2; RING finger protein 79; RNF79 | |
| Recombinant human RFPL2 (immunogen range is 51-150). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| RFPL2 |
| K06336_6A2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| O75678 | |
| 10739 | |
| IgG1 | |
| Protein A | |
| Antibody |