missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RFPL2 Monoclonal Antibody (K06336_6A2), Invitrogen™

Catalog No. MA556036
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556036 200 μL
MA555656 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556036 Supplier Invitrogen™ Supplier No. MA556036
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: DPVGGGNLTN KRPSCAPSPQ DLSAQWKQLE DRGASSRRVD MAALFQEASS CPVCSDYLEK PMSLECGCAV CLKCINSLQK EPHGEDLLCC CSSMVSRKNK

RFPL1, RFPL2 and RFPL3 (ret finger protein-like 1, 2 and 3, respectively), exist as a cluster of genes mapping to human chromosome 22q12.2-q13.3, sharing 95%-96% identity. RFPL1, 2 and 3, are thought to contribute to neocortex organization and size in primates, and show high expression in fetal neocortex as well as embryonic stem-cell neurogenesis. Each of the three RFPL genes encodes two exons giving rise to a putative RING-like motifs and B30-2 domains. RFPL2, or RNF79, is 378 amino acids long and is also highly expressed in prostate with lower expression in fetal kidney and fetal liver. As a result of alternative splicing, two isoforms of RFPL2 exist.

Specifications

Antigen RFPL2
Applications Western Blot
Classification Monoclonal
Clone K06336_6A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene RFPL2
Gene Accession No. O75678
Gene Alias ret finger protein like 2; ret finger protein-like 2; RFPL2; RING finger protein 79; RNF79
Gene Symbols RFPL2
Host Species Mouse
Immunogen Recombinant human RFPL2 (immunogen range is 51-150).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10739
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less