missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RFP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551377 Supplier Invitrogen™ Supplier No. PA551377
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Immunogen sequence: LSRLDTLETS KRKSLQLLTK DSDKVKEFFE KLQHTLDQKK NEILSDFETM KLAVMQAYDP EINKLNTILQ EQRMAFNIAE AFKDVSEPIV FLQQMQEFRE KIKVIKETPL PPSNLPASPL MKNFDTSQWE DIKLVDVDKL SLP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 97%.

The protein encoded by the RFP2 gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies near the nucleus, but its function is unknown. The gene is located on chromosome 13 within the minimal deletion region for B-cell chronic lymphocytic leukemia.

Specifications

Antigen RFP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM13
Gene Accession No. O60858
Gene Alias 3110001L12Rik; B-cell chronic lymphocytic leukemia tumor suppressor Leu5; CAR; CLL-associated RING finger; DLEU5; E3 ubiquitin-protein ligase TRIM13; LEU5; Leukemia-associated protein 5; putative tumor suppressor RFP2; Ret finger protein 2; Rfp2; RING finger protein 77; RING-type E3 ubiquitin transferase TRIM13; RNF77; TRIM13; tripartite motif containing 13; tripartite motif protein 13; tripartite motif-containing 13; tripartite motif-containing protein 13; tripartite motif-containing protein 13; E3 ubiquitin-protein ligase TRIM13; Unknown (protein for MGC:134296)
Gene Symbols TRIM13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RFP2. Recombinant protein control fragment (Product #RP-94749).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10206
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less