missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104384
Description
Antibody detects endogenous levels of total RFC2.
This gene encodes a member of the activator 1 small subunits family. The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins, proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). Replication factor C, also called activator 1, is a protein complex consisting of five distinct subunits. This gene encodes the 40 kD subunit, which has been shown to be responsible for binding ATP and may help promote cell survival. Disruption of this gene is associated with Williams syndrome. Alternatively spliced transcript variants encoding distinct isoforms have been described. A pseudogene of this gene has been defined on chromosome 2.Specifications
| RFC2 | |
| Polyclonal | |
| Unconjugated | |
| RFC2 | |
| 2610008M13Rik; 40kDa; A1 40 kDa subunit; Activator 1 40 kDa subunit; activator 1 subunit 2; Activator 1 subunit C2; AI326953; MGC3665; Recc2; replication factor C (activator 1) 2; replication factor C (activator 1) 2, 40kDa; replication factor C 40 kDa subunit; Replication factor C subunit 2; replication factor C, 40 kDa; RF-C 40 kDa subunit; RFC2; RFC40 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116468, 19718, 5982 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35250, Q641W4, Q9WUK4 | |
| RFC2 | |
| A synthesized peptide derived from human RFC2(Accession P35250), corresponding to amino acid residues K208-P258. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |