missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lewatit NM 60 Ion Exchange Resin (Analytical Grade/Certified), Fisher Chemical
Supplier: Thermo Fisher Scientific R276500
Specifications
| Solid | |
| Analytical Grade | |
| 0.55 mEq/mL min., 1.55mEq/g min. | |
| Poly Bottle | |
| 1.2g/cm³ (340 g/L min. Active Work Density) | |
| 500 g | |
| Certified |
| Lewatit NM 60 ion exchange resin | |
| 69011-49-0 | |
| 16 to 50 Mesh | |
| MFCD00212650 | |
| Brown/Red | |
| Ion Exchange Resin |
Chemical Identifiers
| 69011-49-0 | |
| MFCD00212650 | |
| Ion Exchange Resin |
Safety and Handling