CAS RN 69011-49-0
CAS RN 69011-49-0
IUPAC Name:
(dimethylamino)methanol; 2-[(2-methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate; ethenylbenzene
Synonyms:
(dimethylamino)methanol; glycol dimethacrylate; styrene
Molecular Weight (g/mol):
377.48
Molecular Formula:
C21H31NO5
InChi Key:
RNRCGAPPKMXACN-UHFFFAOYSA-N
SMILES:
CN(C)CO.C=CC1=CC=CC=C1.CC(=C)C(=O)OCCOC(=O)C(C)=C
1
–
1
of
1
results
1
Lewatit NM 60 Ion Exchange Resin (Analytical Grade/Certified), Fisher Chemical
CAS: 69011-49-0 MDL Number: MFCD00212650 Synonym: Ion Exchange Resin