Learn More
Gilson Reversible Tray PIPETMAN™ M96
Reversible tray is a convenient tool for stacking, simplifies pipetting procedures and microplate handling needs
$1528.02 - $4209.06
Specifications
Product Type Tip Holder
Color Aluminium
Quantity 1 Ea.
View More Specs
Products 5
Catalog Number Mfr. No. Includes Positions Price Quantity  
Catalog Number Mfr. No. Includes Positions Price Quantity  
FH07003G
View Documents
Gilson
FH07003
Single Tray 96-384
Each for $1,528.02
Only null left
FH07002G
View Documents
Gilson
FH07002
Rotating Tray 96-96
Each for $1,528.02
Only null left
FH07001G
View Documents
Gilson
FH07001
Rotating Tray 384-384
Each for $1,528.02
Only null left
FH07005G
View Documents
Gilson
FH07005
Rotating Tray 96-384
Each for $4,209.06
Only null left
FH07004G
View Documents
Gilson
FH07004
Multi Tray 96-384
Each for $1,528.02
Only null left
Description
Description
  • The single-tray is a convenient tool for stacking and pipetting in microplates, featuring two sides - one for 96-well microplates and the other for filling a 384-well plate.
  • The reversible rotating tray with 96-96, 384-384 well sides is a convenient accessory for various microplate handling needs.
  • The tray is reversible, suitable for 96-well microplates, and can be fitted on either the left or right side of the PIPETMAN™ M96.
  • The rotating tray with reversible 96 left and 384 right side is a convenient accessory for various microplate handling needs.
  • The tray sides can be flipped to fit either 96- or 384-well microplates and can be positioned on the left or right side of the PIPETMAN™ M96.
  • The PIPETMAN™ M96 multi-tray is a convenient accessory that simplifies pipetting procedures, sliding the tray from one microplate to the next.
  • It is compatible with SBS-standard labware, including 96- and 394-well microplates and reservoirs, and is designed to make tasks such as serial dilutions and plate-to-plate transfers easier.
  • Its sliding mechanism enables efficient movement between positions, optimizing space and saving valuable time during pipetting applications.
  • For increased productivity and minimal plastic waste, the use of PIPETMAN™ DIAMOND Tips Blister Refills and Reload Packs is recommended.
Specifications
Specifications
Tip Holder
Aluminium
1 Ea.