Learn More
Gilson Reversible Tray PIPETMAN™ M96
Reversible tray is a convenient tool for stacking, simplifies pipetting procedures and microplate handling needs
Supplier:  Gilson FH07004
 View more versions of this product
Catalog No. FH07004G
Only null left
Explore available promotions
Description
Description
  • The single-tray is a convenient tool for stacking and pipetting in microplates, featuring two sides - one for 96-well microplates and the other for filling a 384-well plate.
  • The reversible rotating tray with 96-96, 384-384 well sides is a convenient accessory for various microplate handling needs.
  • The tray is reversible, suitable for 96-well microplates, and can be fitted on either the left or right side of the PIPETMAN™ M96.
  • The rotating tray with reversible 96 left and 384 right side is a convenient accessory for various microplate handling needs.
  • The tray sides can be flipped to fit either 96- or 384-well microplates and can be positioned on the left or right side of the PIPETMAN™ M96.
  • The PIPETMAN™ M96 multi-tray is a convenient accessory that simplifies pipetting procedures, sliding the tray from one microplate to the next.
  • It is compatible with SBS-standard labware, including 96- and 394-well microplates and reservoirs, and is designed to make tasks such as serial dilutions and plate-to-plate transfers easier.
  • Its sliding mechanism enables efficient movement between positions, optimizing space and saving valuable time during pipetting applications.
  • For increased productivity and minimal plastic waste, the use of PIPETMAN™ DIAMOND Tips Blister Refills and Reload Packs is recommended.
Specifications
Specifications
Tip Holder
Multi Tray
1 Ea.
0.09 kg
Aluminium
96-384
0.2 lb.
Product Suggestions
Product Suggestions