missing translation for 'onlineSavingsMsg'
Learn More

Reversed Phase Test Mix 2, Certified Reference Material, in Acetonitrile: Water (varied conc.), MilliporeSigma™ Supelco™

Catalog No. 111017238
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017238 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017238 Supplier Merck Emd Millipore Supplier No. 47641U
Only null left

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
Product Line TraceCERT
Physical Form Solution
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) N,N-Diethyl-m-toluamide 600 μg/mL; Phenol 700 μg/mL; Toluene 4000 μg/mL; Uracil 5 μg/mL

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC