missing translation for 'onlineSavingsMsg'
Learn More

G-Biosciences Reverse Transcriptase (RT)

Catalog No. S03143
Encompass
Change view
Click to view available options
No. of Reactions:
10,000
50,000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03143 10,000
S03142 50,000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03143 Supplier G-Biosciences Supplier No. 7861974
Only null left

Reverse Transcriptase is used for first strand cDNA synthesis and gene expression analysis.

  • Reverse Transcriptase (RT) is a recombinant Moloney Murine Leukemia Virus (MMLV) reverse transcriptase with reduced RNase H activity.
  • This RT is an RNA-directed DNA polymerase which lacks 3' - 5' exonuclease activity and can produce full-length cDNA from long RNA templates.

Specifications

Detection Method qPCR
Form Liquid
GC-Rich PCR Performance Medium
PCR Method qPCR, Multiplex qPCR, 1-step RT-qPCR, 2-step RT-qPCR, 1-step RT-PCR
Reaction Speed Fast or Standard
Label or Dye Biotin, FAM, SYBR Green, ROX, TAMRA
Product Type Reverse Transcriptase
For Use With (Application) Gene Expression, Microbial Detection, Pathogen Detection, Virus Detection, cDNA Synthesis
No. of Reactions 10,000