missing translation for 'onlineSavingsMsg'
Learn More

Black Stallion™ Premium Side Split Cowhide Leather Welding Gloves with Aluminized Back

Catalog No. 19050589
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19050589 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19050589 Supplier Black Stallion™ Supplier No. 104XL

missing translation for 'validMainframeMsg'

Welding gloves are constructed of premium quality side-split cowhide leather with an aluminized back. Gloves are wool lined and sewn with Aramid Fiber for additional heat resistance.

  • Premium quality side split cowhide leather
  • Sewn with Aramid Fiber for additional heat resistance
  • Aluminized back
  • Wool lined

Specifications

Back Material Aluminum
Color Orange, Silver
Material Side Split Cowhide Leather
Size X-Large
For Use With (Application) Welding
Glove Cuff Style Safety
Series 104