missing translation for 'onlineSavingsMsg'
Learn More

Black Stallion™ Side Split Cowhide Leather Welder's Gloves with CushionCore™ Liner

CushionCore™ padded liner provides heat-blocking comfort, while Aramid Fiber stitching protects against sparks and abrasion.

$28.59 - $28.59

Specifications

Color Blue, Gray
Hand Specific
Material Side Split Cowhide Leather
For Use With (Application) Stick Welding
Glove Cuff Style Gauntlet
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
28-707-15
View Documents
Black Stallion™
320XL
X-Large
Pair for $28.59
Pack of 12 Pair for $332.28
Only null left
 
Description

Description

  • Side split cowhide leather construction is durable and flexible
  • CushionCore™ padded liner provides heat-blocking comfort
  • Aramid Fiber stitching for spark and abrasion resistance
  • Wide straps on palm for additional reinforcement in a key stress area
  • Welted seams ensure maximum protection for threads
Specifications

Specifications

Blue, Gray
Side Split Cowhide Leather
Gauntlet
Specific
Stick Welding
320
Videos
SDS
Documents

Documents