missing translation for 'onlineSavingsMsg'
Learn More

Black Stallion™ Side Split Cowhide Leather Welder's Gloves with CushionCore™ Liner

Catalog No. 2870715
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
2870715 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2870715 Supplier Black Stallion™ Supplier No. 320 XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CushionCore™ padded liner provides heat-blocking comfort, while Aramid Fiber stitching protects against sparks and abrasion.

  • Side split cowhide leather construction is durable and flexible
  • CushionCore™ padded liner provides heat-blocking comfort
  • Aramid Fiber stitching for spark and abrasion resistance
  • Wide straps on palm for additional reinforcement in a key stress area
  • Welted seams ensure maximum protection for threads

Specifications

Color Blue, Gray
Hand Specific
Material Side Split Cowhide Leather
Size X-Large
For Use With (Application) Stick Welding
Glove Cuff Style Gauntlet
Series 320