Learn More
Bel Art™ Reusable Silica Gel Desiccant Bag
Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators.
$47.58 - $170.47
Specifications
Product Type Desiccant Bag
Description Durable Microfiber Pouch
Material Silica Gel
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
01259109
View Documents Promotion Details
Bel Art™
F420450350
350 g
Each for $47.58
Only null left
01259110
View Documents Promotion Details
Bel Art™
F420450500
500 g
Each for $104.38
Only null left
01259111
View Documents Promotion Details
Bel Art™
F420451000
1000 g
Each for $170.47
Only null left
Description
Description
  • Durable microfiber bag securely contains beads eliminating the risk of leakage and is unaffected by moisture
  • Indicator dot changes from blue when dry to pink when moist to easily identify when recharging is needed
  • Saturated gel can be regenerated by microwaving bag for 6 minutes
  • May be regenerated repeatedly without degradation
  • Arrives individually sealed in moisture-proof pouch
  • Available in multiple sizes to fit most desiccators
Specifications
Specifications
Desiccant Bag
Durable Microfiber Pouch
Silica Gel