Learn More
Corning™ PYREX™ Reusable Heavy-Wall Ignition Tubes
For high-temperature work in the classroom
Supplier:  Corning™ 986014
 View more versions of this product
Catalog No. 14960B
Only null left
Description
Description
These new culture tubes with flat bottoms are ideal for plant tissue culture and general culture work. The flat bottom facilitates use with agar slants. Walls and bottoms are of uniform thickness. The tubes are rimless for greater convenience in plugging and rack storage. Optional polypropylene two position closure. Position closure open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination.
For closures, see Cat. Nos 26796-38, 26797-38, 26794-25, 26795-25, 26798-40, and 26799-40.
Specifications
Chemical Identifiers
Plain
For high-temperature work in the classroom
No
Borosilicate Glass
Non-sterile
Culture Tube
Specifications
Plain
Borosilicate Glass
Non-sterile
14 mm
100 mm
For high-temperature work in the classroom
No
Culture Tube
Product Suggestions
Product Suggestions