missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Reusable Borosilicate Glass Tubes with Plain End

All sizes except 6, 22, and 25mm O.D. have marking spot

Supplier:  Corning™ 982022

 View more versions of this product

Catalog No. 14-957K


Only null left

Description

Description

All sizes except 6, 22, and 25mm O.D. have marking spot

Rimless for greater convenience in plugging and rack storage. Walls and bottoms are of uniform ­thickness. Ends are fire-polished.

Cat. No 9820-12 is a Kahn tubes.

Cat. No. 9820-13 is a Wasserman tube.

Specifications

Specifications

Plain Rimless
175 mm
Non-sterile
22 mm
50 mL
Borosilicate Glass
No
Culture Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions