Learn More
RETSCH ZM200 Ultra Centrifugal Mill: Ring Sieves
Catalog No. 08418144
Change view
Click to view available options
Material:
Stainless Steel
Wear-resistant Stainless Steel
Pore Size:
0.003 in. (0.08 mm)
0.01 in. (0.50 mm)
0.029 in. (0.75 mm)
0.03 in. (1.0 mm)
0.07 in. (2.0 mm)
0.11 in. (3.0 mm)
0.23 in. (6.0 mm)
0.08 mm
0.12 mm
0.20 mm
0.25 mm
0.75 mm
1.0 mm
1.50 mm
2.0 mm
2.00 mm
3.0 mm
3.00 mm
4.0 mm
5.0 mm
5.00 mm
6.0 mm
Pore Style:
Conidur holes
Round holes
Trapezoid holes
Product Type:
Ring Sieve
Ring Sieve with Reinforced Rim
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Pore Size Pore Style Product Type
08418144 Wear-resistant Stainless Steel 0.25 mm Trapezoid holes Ring Sieve with Reinforced Rim
036470235 Stainless Steel 0.01 in. (0.50 mm) Conidur holes Ring Sieve
08418137 Stainless Steel 1.50 mm Trapezoid holes Ring Sieve with Reinforced Rim
08418123 Stainless Steel 1.0 mm Trapezoid holes Ring Sieve
08418134 Stainless Steel 0.01 in. (0.50 mm) Conidur holes Ring Sieve with Reinforced Rim
08418118 Stainless Steel 0.12 mm Trapezoid holes Ring Sieve
08418119 Stainless Steel 0.20 mm Trapezoid holes Ring Sieve
08418117 Stainless Steel 0.08 mm Trapezoid holes Ring Sieve
08418122 Stainless Steel 0.75 mm Trapezoid holes Ring Sieve
08418125 Stainless Steel 2.00 mm Trapezoid holes Ring Sieve
08418127 Stainless Steel 4.0 mm Round holes Ring Sieve
08418124 Stainless Steel 1.50 mm Trapezoid holes Ring Sieve
08418120 Stainless Steel 0.25 mm Trapezoid holes Ring Sieve
08418126 Stainless Steel 3.00 mm Round holes Ring Sieve
08418128 Stainless Steel 5.0 mm Round holes Ring Sieve
08418135 Stainless Steel 0.029 in. (0.75 mm) Conidur holes Ring Sieve with Reinforced Rim
08418130 Stainless Steel 0.003 in. (0.08 mm) Conidur holes Ring Sieve with Reinforced Rim
08418129 Stainless Steel 6.0 mm Round holes Ring Sieve
08418131 Stainless Steel 0.12 mm Trapezoid holes Ring Sieve with Reinforced Rim
08418132 Stainless Steel 0.20 mm Trapezoid holes Ring Sieve with Reinforced Rim
08418138 Stainless Steel 0.07 in. (2.0 mm) Conidur holes Ring Sieve with Reinforced Rim
08418136 Stainless Steel 0.03 in. (1.0 mm) Conidur holes Ring Sieve with Reinforced Rim
08418139 Stainless Steel 0.11 in. (3.0 mm) Round holes Ring Sieve with Reinforced Rim
08418140 Stainless Steel 4.0 mm Round holes Ring Sieve with Reinforced Rim
08418142 Stainless Steel 0.23 in. (6.0 mm) Round holes Ring Sieve with Reinforced Rim
08418141 Stainless Steel 5.00 mm Round holes Ring Sieve with Reinforced Rim
08418145 Stainless Steel 0.01 in. (0.50 mm) Conidur holes Ring Sieve with Reinforced Rim
08418143 Wear-resistant Stainless Steel 0.20 mm Trapezoid holes Ring Sieve with Reinforced Rim
08418146 Wear-resistant Stainless Steel 0.75 mm Trapezoid holes Ring Sieve with Reinforced Rim
08418149 Wear-resistant Stainless Steel 2.0 mm Trapezoid holes Ring Sieve with Reinforced Rim
08418147 Wear-resistant Stainless Steel 1.0 mm Trapezoid holes Ring Sieve with Reinforced Rim
08418150 Wear-resistant Stainless Steel 3.0 mm Round holes Ring Sieve with Reinforced Rim
08418148 Wear-resistant Stainless Steel 1.50 mm Trapezoid holes Ring Sieve with Reinforced Rim
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
Catalog No. 08418144 Supplier RETSCH Supplier No. 036470263
Only null left
Stainless-steel sieves for use with ZM200 Ultra Centrifugal Mill
Specifications
Product Type Ring Sieve with Reinforced Rim
For Use With (Equipment) ZM200 ultra centrifugal mil
Material Wear-resistant Stainless Steel
Pore Size 0.25 mm
Pore Style Trapezoid holes