Learn More
RETSCH ZM 200 Ultra Centrifugal Mill, Push-Fit Rotors
For fine grinding of soft to medium-hard and fibrous materials.
Supplier: RETSCH 026080041
Description
- Accessory for use with ZM 200 Ultra Centrifugal Mill
- Wear resistant coating
Specifications
| Stainless Steel | |
| 6,000 to 18,000 rpm | |
| Centrifugal Mill |
| 12 Tooth SS - Medium Coarse Materials | |
| 9.9 cm |