Learn More
RETSCH PM 100 Planterary Ball Mill Grinding Balls
Catalog No. 08418187
Change view
Click to view available options
Material:
Agate
Stainless Steel
Zirconium Oxide
Diameter (Metric):
0.2 cm
0.3 cm
1 cm
2 cm
For Use With (Application):
Ideal for colloidal milling
Ideal for standard grinding
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Diameter (Metric) For Use With (Application)
08418187 Zirconium Oxide 1 cm Ideal for standard grinding
08418182 Stainless Steel 0.3 cm Ideal for colloidal milling
053680028 Agate 2 cm Ideal for standard grinding
08418181 Stainless Steel 0.2 cm Ideal for colloidal milling
08418177 Stainless Steel 1 cm Ideal for colloidal milling
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 08418187 Supplier RETSCH Supplier No. 053680094
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Retsch PM 100 and PM 200 Planetary Ball Mills
  • In assorted diameters and materials
  • Smaller balls offered for colloidal milling
Specifications
Product Type Grinding Ball
Diameter (English) 0.39 in.
Diameter (Metric) 1 cm
For Use With (Application) Ideal for standard grinding
For Use With (Equipment) PM 100 and PM 200 planetary ball mill
Material Zirconium Oxide