Learn More
RETSCH Knife Mill Grindomix GM300 Accessories: Grinding Containers
Catalog No. 08418327
Change view
Click to view available options
Product Type:
Grinding Container
Grinding Container with Knife Holder
Material:
Plastic
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material
08418327 Grinding Container Plastic
08418326 Grinding Container with Knife Holder Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08418327 Supplier RETSCH Supplier No. 30450056
Accessories for Knife Mill Grindomix GM300.
For use with Knife Mill Grindomix GM300 (Cat. No. 08-418-325).
Specifications
Product Type Grinding Container
Capacity (Metric) 5 L
For Use With (Equipment) For Knife Mill Grindomix GM300
Material Plastic