Learn More
RETSCH CryoMill Grinding Jar
Catalog No. 08418233
Change view
Click to view available options
For Use With (Equipment):
Adapter of CryoMill Grinder
Catalog No. For Use With (Equipment)
08418233 Adapter of CryoMill Grinder
1 options
Catalog No. 08418233 Supplier RETSCH Supplier No. 014620290
Helps provide efficient milling
  • Stainless-steel grinding jar with screw-top design
  • Grinding jar is continually cooled with liquid nitrogen from the integrated cooling system before and during the grinding process
Specifications
Product Type Grinding Jar
Capacity (English) 0.16 oz.
Capacity (Metric) 5 mL
Closure Type Screw Top
For Use With (Application) Ideal for cryogenic grinding
For Use With (Equipment) Adapter of CryoMill Grinder
Material Stainless Steel