Learn More
Kent Scientific Retractor Tip for SurgiSuite Multi-Functional Surgical Platforms
For use with SurgiSuite Multi-Functional Surgical Platforms. Kent Scientific Retractor Tip is locked onto wire for static retraction, or affixed to wire for dynamic retraction.
Supplier:  Kent Scientific SURGI5015
Encompass
 View more versions of this product
Catalog No. 13005190
Description
Description
  • Locked onto wire for static retraction
  • Affixed to elastic wire for dynamic retractor
Specifications
Chemical Identifiers
Surgery, Homeothermic Warming, Surgical Procedures, Tissue Retraction
Retractor Tip
SurgiSuite and SurgiSuite-LG Multi-Functional Surgical Platforms
Specifications
Surgery, Homeothermic Warming, Surgical Procedures, Tissue Retraction
SurgiSuite and SurgiSuite-LG Multi-Functional Surgical Platforms
Blunt
Retractor Tip
1 mm
Product Suggestions
Product Suggestions