missing translation for 'onlineSavingsMsg'
Learn More

Retn, Mouse anti-Rat, Clone: RRES 07, Abnova™

Catalog No. 89998607
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89998607 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89998607 Supplier Abnova Corporation Supplier No. MAB15873
Only null left

Mouse Monoclonal Antibody

Specifications

Antigen Retn
Applications ELISA, Western Blot
Classification Monoclonal
Clone RRES 07
Conjugate Unconjugated
Dilution ELISA (1:5000-1:10000) Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4.
Gene resistin
Gene Alias -
Gene Symbols Retn
Host Species Mouse
Immunogen Recombinant protein corresponding to rat Retn.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246250
Target Species Rat
Content And Storage For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less