Learn More
Thermo Scientific™ Retention Gap Kits
Install a retention gap/guard column to your analytical column with a single kit.
$122.21
Specifications
For Use With Guard Column (GC)
Type Retention Gap Kit
For Use With (Application) GC
View More Specs
Products 1
Catalog Number Mfr. No. Overall Length (Metric) Diameter (Metric) Inner Material Price Quantity & Availability  
Catalog Number Mfr. No. Overall Length (Metric) Diameter (Metric) Inner Material Price Quantity & Availability  
03150233
View Documents Promotion Details
Thermo Scientific™
260G498P
2 m 0.32 mm Deactivated Silica
Each for $122.21
Only null left
Description
Description
Use Thermo Scientific™ Retention Gap Kits to conveniently install a retention gap/guard column to your analytical column. Each kit includes deactivated silica tubing, a miniunion, and five ferrules.
Includes:
  • Length of deactivated fused silica tubing
  • Mini-union
  • Five ferrules
Specifications
Specifications
Guard Column (GC)
Retention Gap Kit
GC
Videos
SDS
Documents
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.