missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Retainer & Base Set (Fast) for 3500/3500xL Genetic Analyzers, 8 tube

Catalog No. 4410233
Encompass
Change view
Click to view available options
Quantity:
4 Sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4410233 4 Sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4410233 Supplier Applied Biosystems™ Supplier No. 4410233

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This 8-tube retainer and base set is for use with Fast 8-tube strips (0.1mL) on the 3500 series Genetic Analyzers.

This 8-tube retainer and base set is for use with Fast 8-tube strips (0.1 ml) on the 3500 series Genetic Analyzers.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Content And Storage This product includes 4 sets of retainers and bases.
Description Retainer & Base Set (Fast), 4 Sets, 8-well Strips, 0.1mL, Strip Tube Type, For 3500/3500xL Genetic Analyzers, Room Temperature
Format 8-well Strip
For Use With (Equipment) 3500 Genetic Analyzer, 3500xL Genetic Analyzer
For Use With (Application) Sequencing
Product Type Base Set
Quantity 4 Sets
Sample Volume 0.1 mL
Shipping Condition Room Temperature
Tube Type Strip Tube
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?

If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.

For Research Use Only. Not for use in diagnostic procedures.