missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Resveratrol | |
| 267°C | |
| C14H12O3 | |
| 25 g | |
| LUKBXSAWLPMMSZ-OWOJBTEDSA-N | |
| 5-[(E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol | |
| 445154 | |
| 228.25 | |
| Crystalline Powder |
| 501-36-0 | |
| Yellow | |
| MFCD00133799 | |
| resveratrol, trans-resveratrol, 3,4',5-trihydroxystilbene, 3,5,4'-trihydroxystilbene, e-resveratrol, 3,4',5-stilbenetriol, e-5-4-hydroxystyryl benzene-1,3-diol, resvida, 3,4',5-trihydroxy-trans-stilbene, 5-e-2-4-hydroxyphenyl ethenyl benzene-1,3-diol | |
| C1=CC(=CC=C1C=CC2=CC(=CC(=C2)O)O)O | |
| 228.247 | |
| CHEBI:45713 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 501-36-0 | |
| 228.247 | |
| LUKBXSAWLPMMSZ-OWOJBTEDSA-N | |
| 445154 | |
| 5-[(E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol |
| C14H12O3 | |
| MFCD00133799 | |
| resveratrol, trans-resveratrol, 3,4',5-trihydroxystilbene, 3,5,4'-trihydroxystilbene, e-resveratrol, 3,4',5-stilbenetriol, e-5-4-hydroxystyryl benzene-1,3-diol, resvida, 3,4',5-trihydroxy-trans-stilbene, 5-e-2-4-hydroxyphenyl ethenyl benzene-1,3-diol | |
| CHEBI:45713 | |
| C1=CC(=CC=C1C=CC2=CC(=CC(=C2)O)O)O |
Safety and Handling
RTECSNumber : CA8987000
TSCA : No
Recommended Storage : Refrigerator