missing translation for 'onlineSavingsMsg'
Learn More

Resveratrol, MilliporeSigma™ Supelco™

Catalog No. 111014345
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014345 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014345 Supplier Merck Emd Millipore Supplier No. 34092100MG
Only null left

Resveratrol, is a naturally occurring phytoalexin, which is produced by spermatophytes like grapevines and mulberries and other food products. It can serve as a potential candidate, which shows potent antiestrogenic activity and hence suppress the growth of cancer cells.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 501-36-0
Physical Form Neat
Linear Formula C14H12O3
MDL Number MFCD00133799
Synonym 3,4 ′,5-Trihydroxy-trans-stilbene; 5-[(1 E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol
Type Analytical Standard
Quantity 100 mg
Formula Weight 228.24
Grade Analytical Standard