missing translation for 'onlineSavingsMsg'
Learn More

Resveratrol, 98%

Catalog No. AAJ60790ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60790ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60790ME Supplier Thermo Scientific Chemicals Supplier No. J60790ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 501-36-0 | C14H12O3 | 228.247 g/mol

It is used as a natural phenolic antioxidant that is a potent activator of SIRT1 and inhibitor of Cox-1. Resveratrol has also been used extensively in stem cell studies to enhance self-renewal and multipotency of mesencymal stem cells (MSC’s) in vitro. Resveratrol acts as a NF-κB inhibitor to protect MSC-derived chondrocytes in vitro. Resveratrol is used to treat adipose-derived mesenchymal stem cells (ADMSCs) resulted in increased alkaline phosphatase activity and osteocalcin levels; thus enriching the MSC and osteoprogenitor cell populations in cultured ADMSCs in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a natural phenolic antioxidant that is a potent activator of SIRT1 and inhibitor of Cox-1. Resveratrol has also been used extensively in stem cell studies to enhance self-renewal and multipotency of mesencymal stem cel

Chemical Identifiers

CAS 501-36-0
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00133799
InChI Key LUKBXSAWLPMMSZ-OWOJBTEDSA-N
Synonym resveratrol, trans-resveratrol, 3,4',5-trihydroxystilbene, 3,5,4'-trihydroxystilbene, e-resveratrol, 3,4',5-stilbenetriol, e-5-4-hydroxystyryl benzene-1,3-diol, resvida, 3,4',5-trihydroxy-trans-stilbene, 5-e-2-4-hydroxyphenyl ethenyl benzene-1,3-diol
PubChem CID 445154
ChEBI CHEBI:45713
IUPAC Name 5-[(E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
SMILES C1=CC(=CC=C1C=CC2=CC(=CC(=C2)O)O)O

Specifications

Color White
Quantity 500 mg
Merck Index 14,8158
Solubility Information Soluble in water (3mg/100mL),ethanol (50mg/mL),DMSO (?16mg/mL),DMF (∽65mg/mL),PBS (pH 7.2) (∽100μg/mL),methanol,and acetone (50mg/mL).
Formula Weight 228.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Resveratrol
Hazard Category H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
RTECSNumber CZ8987000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only