missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Resurge™ CD2 Supplement, without glucose and L-glutamine
GREENER_CHOICE

Catalog No. p-8843006
Change view
Click to view available options
Quantity:
100 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
670015 100 g
670017 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 670015 Supplier Gibco™ Supplier No. 670015
Only null left

A diverse set of fully chemically defined, animal origin–free and protein-free cell culture supplements, which help promote cell growth and boost protein production.

A chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Culture Type Mammalian Cell Culture
Form Powder
Product Type Media Supplement
Without Additives No Glutamine
Format Bottle(s)
Product Line Resurge
Quantity 100 g
Source Animal origin-free
Show More Show Less