missing translation for 'onlineSavingsMsg'
Learn More

Promega Restriction Enzymes - Xho I Blue/White Cloning and Genome Qualified (D)

Catalog No. p-2387589
Encompass
Change view
Click to view available options
Quantity:
10,000 U
3000 U
Concentration:
10 U/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PRR6165 10,000 U 10 U/μL
PRR6161 3000 U 10 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRR6165 Supplier Promega Supplier No. R6165
Only null left

Capable of digesting DNA in 15 minutes or less. Genome qualified to ensure optimal performance in genomic analysis applications. (C/TCGA G G AGCT\C)

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 10 U/μL
Components 1 x 1mL 10X Buffer D,1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 10000U XhoI
Incubator Temperature 37°C
pH 7.3
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Storage Buffer 10mM Tris HCl (pH 7.3), 300mM KCl, 0.1% Triton™ X-100, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 10,000 U
Cut Site C.TCGAG GAGCT.C
Product Type XhoI
Form Liquid
Show More Show Less