missing translation for 'onlineSavingsMsg'
Learn More

Promega Restriction Enzymes - EcoR V Blue/White Cloning Qualified (D) DFS Item

Capable of digesting DNA in 15 minutes or less. (GAT/ATC CTA\TAG)

$81.84 - $308.32

Specifications

Incubator Temperature 37°C
pH 7.4
For Use With (Application) Capable of digesting DNA in <15 min.
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Cut Site GAT.ATC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Concentration Price Quantity  
Catalog Number Mfr. No. Quantity Concentration Price Quantity  
PR-R6355 DFS Item
View Documents
Promega
R6355
10,000 U 10 U/μL
Each for $308.32
Only null left
 
PR-R6351 DFS Item
View Documents
Promega
R6351
2000 U 10 U/μL
Each for $81.84
Only null left
 
Description

Description

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Specifications

37°C
Capable of digesting DNA in <15 min.
GAT.ATC
7.4
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'