missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Restore™ PLUS Western Blot Stripping Buffer

Catalog No. PI46428
Change view
Click to view available options
Quantity:
30 mL
500 mL
4 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI46428 30 mL
FER46430X4 4 x 500 mL
PI46430 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI46428 Supplier Thermo Scientific™ Supplier No. 46428

Remove high-affinity antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.

Thermo Scientific Restore PLUS Western Blot Stripping Buffer is an advanced formula for removing bound primary and secondary antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.

Features of Restore PLUS Western Blot Stripping Buffer:

  • Ready and easy to use—no dilution necessary; no offensive odors; store at room temperature
  • Compatible—use on nitrocellulose and PVDF membranes, whether still wet or already dry; works with practically any blocking buffer, enzyme conjugate and chemiluminescent substrate
  • Cost effective—save valuable time and samples; strip blots effectively the first time
  • Robust yet gentle—transferred proteins remain viable; strip the same blot up to five times
  • Flexible—strip and reprobe to optimize antibody concentrations or to detect a new antigen with different antibodies

Restore PLUS Buffer is an alternative formulation of the original Thermo Scientific Restore Western Blot Stripping Buffer. Restore PLUS Stripping Buffer was designed for use with antibodies that are difficult to remove from western blots and require longer incubation times or incubation temperatures greater than 22°C with gentler formulations. High-affinity antibodies can be quickly and effectively stripped from western blots at room temperature without removing transferred proteins, thereby allowing multiple reprobes of the target.

Protocol Summary:

  • Wash blot to remove chemiluminescent substrate
  • Incubate blot in Restore Western Blot Stripping Buffer for 5 to 15 minutes at room temperature
    • (or incubate at 37°C for high affinity antibodies)
  • Remove blot and wash in Wash Buffer
  • Block membrane
  • Test for sufficient removal of antibodies
  • Perform next immunoblot experiment

Specifications

Product Type Western Blot Stripping Buffer
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line Restore
Quantity 30 mL
Content And Storage Upon receipt store at room temperature. Product shipped at ambient temperature.
How many times can I strip my blot using Restore or Restore Plus Western Blot Stripping Buffers ?

The stability of the attached (transferred and bound) protein will determine the number of times the membrane can be successfully re-probed after stripping. The protein may withstand stripping as many as four times or as few as one time.

What is the proper wash buffer to use after stripping with Restore or Restore Plus Western Blot Stripping buffers?

Wash the membrane with the same buffer as was used between antibody probing steps during the Western blotting procedure.

Can I strip membranes with Restore or Restore Plus Western Blot Stripping Buffer and reprobe for subsequent detection with fluorescent probes?

No. Although the fluorescent antibodies, like other antibodies, can be stripped with Restore Buffers, stripped membranes typically produce unacceptable background for subsequent fluorescent detection methods. We recommend Restore Fluorescent Western Blot Stripping Buffer, Cat. No. 62299 (20 mL) and Cat. No. 62300 (100 mL).

Note: Restore Fluorescent Western Blot Stripping Buffer is for use with low-fluorescence PVDF membrane (Cat. No. 22860) only.

Will Restore and Restore Plus Western Blot Stripping Buffers remove biotin-bound avidin, streptavidin or NeutrAvidin Protein conjugates?

No. Restore and Restore Plus Buffers are not stringent enough to break the avidin-biotin interaction. A buffer capable of breaking this interaction would likely also damage the target protein, making it undetectable. However it may be possible to strip the complexed biotinylated primary antibody and streptavidin-HRP secondary antibody from the target protein on the membrane. Please also see Tech Tip: Strip and Reprobe Western Blots (https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/strip-reprobe-western-blots-tech-tip.pdf).

Will the Restore and Restore Plus Western Blot Stripping Buffers remove precipitating substrates?

No. The antibodies are removed but the substrate leaves a permanent precipitate on the membrane that cannot be removed. Restore and Restore Plus Buffers are designed for procedures using chemiluminescent substrates. Please note that this is not compatible with fluorescence supstrates as it will result in increased background. For fluorescent substrates please use our Restore Fluorescent Western Blot Stripping Buffer. Please also see Tech Tip: Strip and Reprobe Western Blots (https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/strip-reprobe-western-blots-tech-tip.pdf).

Do I need to re-block the membrane after stripping with Restore and Restore Plus Western Blot Stripping Buffers?

After stripping with Restore Buffer, re-blocking of the membrane is usually not necessary but may help to decrease background in some situations. By contrast, re-blocking is required after stripping with Restore Plus Buffer.

Can Restore and Restore Plus Western Blot Stripping Buffers be used on both nitrocellulose and PVDF membranes?

Yes. The stripping buffers work to separate the antibody from the antigen, so the membrane to which the antigen is bound generally will not affect the stripping.

Do I need to re-optimize conditions when switching from Restore to Restore Plus Western Blot Stripping Buffer?

Yes. Both incubation time and temperature must be optimized for best results.

Have the Restore and Restore Plus Western Blot Stripping Buffers been tested for my primary or secondary antibody?

No. Every binding interaction is slightly different in terms of mode (i.e., actual amino acids or functional groups that interact) and affinity (strength of binding under a given set of buffer conditions). First try Restore Western Blot Stripping Buffer for 15 minutes at room temperature. If antibody removal is incomplete, optimize the stripping conditions by increasing the time and temperature. If this fails to completely strip the antibodies, then switch to the more stringent Restore Plus Western Blot Stripping Buffer.

What is the difference between the Restore and Restore Plus Western Blot Stripping Buffers?

Restore Western Blot Stripping Buffer gently but effectively removes the primary and secondary antibodies from the membrane to allow re-probing on the same membrane. The buffer is formulated to work for a wide variety of interactions, but there are some high affinity antigen-antibody interactions that require more stringent stripping conditions. Restore Plus Western Blot Stripping Buffer was developed for these difficult to strip interactions.

What Western blot stripping buffers do you offer?

There are two types of stripping buffers, each available in two different sizes:

• Restore Western Blot Stripping Buffer: Cat. No. 21059 (500 mL), Cat. No. 21063 (5 L) and Cat. No. 21062 (30 mL)
• Restore Plus Western Blot Stripping Buffer: Cat. No. 46430 (500 mL) and Cat. No. 46428 (30 mL)

If I want to strip and re-probe a nitrocellulose membrane, what procedure do you recommend?

Thermo Fisher Scientific sells 4 ready-to-use buffers specifically intended for stripping Western blots prior to re-probing. Three of them can be used with both nitocellulose and PVDF membranes. We have Restore Western Blot Stripping Buffer (Cat. No. 21059), a trial size of this product (Cat. No. 21062), a product called Restore PLUS (Cat. No. 46428), which is formulated to strip off antibodies that are difficult to remove, and Restore Fluorescent Western Blot Stripping Buffer (Cat. No. 62299). The latter is used to strip fluorophore-labeled antibodies from PVDF membranes only.

How can I store, strip, and reuse my western blot?

For nitrocellulose or PVDF membrane following Western blot detection using a chemiluminescent or fluorescent substrate system: Following transfer, air dry the membrane and place in an envelope, preferably on top of a supported surface to keep the membrane flat. The blot can be stored indefinitely at -80 degrees C. When ready to reprobe, prewet the PVDF blot with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with blocking step.

FOR STRIPPING/REPROBING OF MEMBRANES: Harsh protocol (see NOTE below for modifications)

1) Submerge the membrane in stripping buffer (100 mM BME, 2% SDS, 62.5 mM Tris-HCl, pH 6.7) and incubate at 50 degrees C for 30 min with occasional agitation. If more stringent conditions necessary, incubate at 70 degrees C.

2) Wash 2 x 10 min in TBS-T/PBS-T at room temperature.

3) Block the membrane by immersing in 5% blocking reagent TBS-T or PBS-T for 1 hr at room temperature.

4) Immunodetection

NOTE: Often you don't need such harsh conditions to remove antibodies from their proteins. The stringency of one or several of the variables can be decreased: lower the temperature, decrease the time, less BME, less SDS, etc. An especially mild but still often effective stripping protocol is lower pH incubation. Example: pH 2.0 Tris 50-100 mM, 30-60 min incubation (you may do two incubations if you wish). Then rinse and block as usual. If you do not wish to re-use the membrane immediately after stripping, you can store the membrane in plastic wrap (wet, you do not want it to dry out). Another simple, mild stripping buffer is 0.1 M glycine•HCl (pH 2.5-3.0), incubation 30 min to 2 hrs room temperature or 37 degrees C, depending on the antibody.


For Research Use Only. Not for use in diagnostic procedures.