missing translation for 'onlineSavingsMsg'
Learn More

Restek™ TopLoader BalanceBank Storage Unit

Catalog No. 06712704
Encompass
Change view
Click to view available options
Product Type:
Storage Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
06712704 Storage Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06712704 Supplier Restek™ Supplier No. RK 25148

Creates a neat, highly functional weighing area and workstation. Restek™ TopLoader BalanceBank Storage Unit provides easy access to wipers, weigh papers, boats, and spatulas.

  • Provides easy access to wipers, weigh papers, boats, and spatulas
  • Creates a neat, highly functional weighing area and workstation

Specifications

Product Type Storage Unit
Dimensions (L x W x H) 30.4 x 30.4 x 17.7 cm (12 x 12 x 7 in.)
For Use With (Equipment) Wipers, weigh Papers, boats and spatulas
For Use With (Application) Provides easy access, creates a neat, highly functional weighing area and workstation