missing translation for 'onlineSavingsMsg'
Learn More

Restek™ TO-14A Internal Standard/Tuning Mix

Catalog No. 06802379
Encompass
Change view
Click to view available options
Concentration:
1 ppm
100 particles/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
06802379 1 ppm
06802390 100 particles/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06802379 Supplier Restek™ Supplier No. RK 34408

Includes

Certificate of Analysis and unique serial number

Cylinders are disposable and do not require rental or demurrage fees

Calibration standards are provided by Spectra/Linde.

  • Meets lab requirements for two separate sources of calibration standards
  • Minimum 12-month stability on all cylinders
  • Aluminum construction
  • Size: 8 × 24cm
  • Volume/Pressure: 104L of gas at 1800 psi
  • CGA-180 outlet fitting
  • Weight: 0.7kg (1.5 lb.)

Compounds

  • Bromochloromethane
  • 1-Bromo-4-fluorobenzene (4-Bromofluorobenzene)
  • Chlorobenzene-d5
  • 1,4-Difluorobenzene

Order Info

Gas standards are subject to hazardous materials shipping fees by most freight carriers. All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements.

Specifications

Components Bromochloromethane, 4-Bromofluorobenzene, Chlorobenzene-d5, 1,4-Difluorobenzene in Nitrogen
Concentration 1 ppm
Volume (Metric) Gas 104 L
Pressure 1800 psi
Weight (English) 1.5 lb.
Volume (English) Gas 27.47 gal.
Weight (Metric) 0.7 kg
Type TO-14A Internal Standard/Tuning Mix