Learn More
Restek™ Syringe Adapter Kit
For Single-Stage VOC Regulator
$183.25
Specifications
| For Use With (Application) | Use to withdraw sample from a high pressure cylinder after pressure reduction through the high purity VOC single stage regulator |
|---|---|
| Includes | One Nickel-plated Brass 0.25 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, one Stainless Steel, 0.25 in. NPT x 0.125 in. compression fitting with septum (can be used with any syringe needle) |
Includes: One nickel-plated brass 1/4 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, One stainless steel, 1/4 in. NPT x 1/8 in. compression fitting with septum (can be used with any syringe needle).
Description
- Use to withdraw sample from a high-pressure cylinder after pressure reduction through the high-purity VOC single-stage regulator
Specifications
| Use to withdraw sample from a high pressure cylinder after pressure reduction through the high purity VOC single stage regulator | |
| One Nickel-plated Brass 0.25 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, one Stainless Steel, 0.25 in. NPT x 0.125 in. compression fitting with septum (can be used with any syringe needle) |