Learn More
Restek™ Stainless-Steel HPLC Column Tubing
Can be used immediately without any additional treatment
Supplier: Restek™ RK25106
Description
- Manufactured from fine, chromatographic-grade Type 316 stainless steel
- Corrosion-resistant, ultrasonically cleaned and passivated
- Tubing is polished to ensure a burr-free cut
Specifications
| HPLC Column Tubing | |
| 0.25 in. | |
| 1.2 in. | |
| 316 Stainless Steel |
| 0.125 in. | |
| 0.635 cm | |
| 3 cm |