Learn More
Restek™ Resprep™-C18 and Resprep™-C8 SPE Disks
Extract analytes using high flow rates with significantly less clogging
$891.32 - $897.38
Specifications
Certifications/Compliance EPA 525.1, 506, 550.1, 549.1
Type SPE Disk
Material Glass Fiber
Diameter (Metric) Outer 4.7 cm
View More Specs
Products 2
Catalog Number Mfr. No. Sorbent Price Quantity & Availability  
Catalog Number Mfr. No. Sorbent Price Quantity & Availability  
06712142
Encompass
View Documents Promotion Details
Restek™
RK 24004
C18
Each for $897.38
Only null left
06718826
Encompass
View Documents Promotion Details
Restek™
RK 24048
C8
Pack of 24 Each for $891.32
Only null left
Description
Description
  • Glass fiber disks embedded with C18 or C8 bonded silica
  • Extract semivolatile organic compounds
  • Deep-pore design reduces clogging and allows faster flow rates
  • Meet requirements for EPA methods 525.1, 506, 550.1, 549.1
Specifications
Specifications
EPA 525.1, 506, 550.1, 549.1
SPE Disk
Glass Fiber
4.7 cm