Learn More
Restek™ Resprep™-C18 and Resprep™-C8 SPE Disks
Extract analytes using high flow rates with significantly less clogging
Supplier: Restek™ RK 24048
Description
- Glass fiber disks embedded with C18 or C8 bonded silica
- Extract semivolatile organic compounds
- Deep-pore design reduces clogging and allows faster flow rates
- Meet requirements for EPA methods 525.1, 506, 550.1, 549.1
Chemical Identifiers
| EPA 525.1, 506, 550.1, 549.1 | |
| SPE Disk | |
| Glass Fiber | |
| 4.7 cm |
Specifications
| EPA 525.1, 506, 550.1, 549.1 | |
| SPE Disk | |
| 4.7 cm |
| C8 | |
| Glass Fiber |