missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Replacement Inlet Seals for Agilent 5890/6890 Split/Splitless Injection Ports DFS Item

For use with Agilent GCs.

$230.57 - $234.45

Specifications

Type Seal
Diameter (Metric) Inner 1.2 mm
Packaging Quantity 2 Pack
For Use With Agilent 5890/6890 Split/Splitless Injection Ports
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
06718489 DFS Item
View Documents
Restek™
RK21307
Siltek N/A
Only null left
 
06718487 DFS Item
View Documents
Restek™
RK21305
Gold-plated N/A
Only null left
 
Description

Description

  • Siltek treatment provides enhanced inertness versus stainless steel
  • Include washers
  • Available Siltek or gold-plated seals
  • I.D.: 1.2mm

Note
The 1.2mm inlet seal is recommended when installing two columns using a two-hole Vespel/graphite ferrule.

Specifications

Specifications

Seal
1.2 mm
2 Pack
Agilent 5890/6890 Split/Splitless Injection Ports
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'