missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Replacement Inlet Seals for Agilent 5890/6890 Split/Splitless Injection Ports DFS Item

For use with Agilent GCs.

Supplier:  Restek™ RK21305

 View more versions of this product

Catalog No. 06718487


Only null left
Explore available promotions

Description

Description

  • Siltek treatment provides enhanced inertness versus stainless steel
  • Include washers
  • Available Siltek or gold-plated seals
  • I.D.: 1.2mm

Note
The 1.2mm inlet seal is recommended when installing two columns using a two-hole Vespel/graphite ferrule.

Specifications

Specifications

Seal
2 Pack
Gold-plated
1.2 mm
Agilent 5890/6890 Split/Splitless Injection Ports
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions