Learn More
Restek™ Polypropylene Vial Inserts
Specially designed for 2.0mL, 11mm crimp-top and 2.0mL, 9mm short-cap, screw-thread vials.
Supplier: Restek™ RK 24520
Description
- Big mouth
- Pack of 100
Chemical Identifiers
| Polypropylene | |
| 100/Pk. | |
| Vial Insert |
Specifications
| Polypropylene | |
| Big Mouth, No Spring | |
| Vial Insert |
| 250 μL | |
| 100/Pk. |