Learn More
Restek™ MXT Y-Union Connector Kits For Fused Silica Columns DFS Item
For connecting metal and/or fused silica GC columns.
Supplier:  Restek™ RK21387
 View more versions of this product
Catalog No. 06711569
Only null left
Explore available promotions
Description
Description
These MXT connectors can be used with fused silica tubing because of a Valcon polyimide, 1/32 in., one-piece fused silica adapter. This unique graphite-reinforced composite allows a capillary column to slide into the adapter and be locked in place simply by loosening and tightening the fitting.
  • Low-dead-volume, leak-tight connection
  • Reusable
  • Siltek treatment ensures maximum inertness
  • Ideal for connecting a guard column or transfer line to an analytical column
  • Use to oven temperatures of 350°C
  • Can also be used for connecting fused silica to metal
  • Includes MXT union, three 1/32 in. nuts and three one-piece, fused silica adapters
Specifications
Specifications
Connector
0.53 mm
MXT union, three 1/32 in. nuts and three one-piece, fused silica adapters
0.53 mm I.D. Fused Silica Columns
Product Suggestions
Product Suggestions