Learn More
Restek™ Micropacked GC Columns: Mole Sieve DFS Item
Efficient, inert, and flexible micropacked columns are highly efficient and provide good sample capacity, resulting in a powerful tool for solving many difficult application problems.
$565.87 - $726.64
Specifications
For Use With (Application) GC
Material Stainless Steel
Mesh Size 80/100
View More Specs
Products 8
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Outer Packing Material Max. Temperature Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Outer Packing Material Max. Temperature Price Quantity  
06710371 DFS Item
View Documents
Restek™
RK19001
2 m 1.59 mm Mole Sieve 5Å 300°C
Each for $695.26
Only null left
06710370 DFS Item
View Documents
Restek™
RK19000
1 m 1.59 mm Mole Sieve 5Å 300°C
Each for $565.87
Only null left
06710372 DFS Item
View Documents
Restek™
RK19002
1 m 0.95 mm Mole Sieve 5Å 300°C
Each for $581.87
Only null left
06710376 DFS Item
View Documents
Restek™
RK19005
2 m 1.59 mm Mole Sieve 13X 350°C
Each for $726.64
Only null left
06710375 DFS Item
View Documents
Restek™
RK19004
1 m 1.59 mm Mole Sieve 13X 350°C
Each for $576.49
Only null left
06710374 DFS Item
View Documents
Restek™
RK19003
2 m 0.95 mm Mole Sieve 5Å 300°C
Each for $724.64
Only null left
06710378 DFS Item
View Documents
Restek™
RK19006
1 m 0.95 mm Mole Sieve 13X 350°C
Each for $576.49
Only null left
06710379 DFS Item
View Documents
Restek™
RK19007
2 m 0.95 mm Mole Sieve 13X 350°C
Each for $726.64
Only null left
Description
Description
  • Increased efficiency over traditional packed columns
  • Higher capacity than PLOT columns
  • Made from inert, flexible, SilcoSmooth tubing
  • Standard coils fit all instruments; no special instrument configurations required
Efficient, Inert, and Flexible
  • Unsurpassed inertness of SilcoSmooth tubing is based on Siltek deactivation
  • This allows the column to be flexed and coiled without any fear of chipping or cracking the inert surface
Easy to Install - Multiple Internal Diameters
  • Designed to fit packed and capillary injection systems
  • Standard wall (1/16 in. OD) micropacked columns offer improved efficiency in packed column instruments without the expense of converting to capillary injection systems
  • Smaller OD (0.95mm OD) micropacked columns install easily into a capillary injector, using slightly larger ferrules
  • Operate at flows exceeding 10 cc/min. for trouble-free operation
Braided Wire End Plugs
  • Braided wires are inserted into the column and secured by making a small crimp near the column outlet
  • End plugs are Siltek treated which makes the sample contacts only inert surfaces
  • Mesh Size: 80/100
Order Info
Custom packed and micropacked columns also available by request.
Specifications
Specifications
GC
Stainless Steel
80/100
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'