missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Micropacked GC Columns: Hayesep Q DFS Item

Efficient, inert, and flexible micropacked columns are highly efficient and provide good sample capacity, resulting in a powerful tool for solving many difficult application problems.

$623.75 - $783.64

Specifications

Max. Temperature 275°C
For Use With (Application) GC
Material Stainless Steel
Packing Material HayeSep Q
Mesh Size 100/120
View More Specs

Products 4
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity  
06710389 DFS Item
View Documents
Restek™
RK19016
1 m 1 mm 1.59 mm
Each for $623.75
Only null left
 
06710390 DFS Item
View Documents
Restek™
RK19017
2 m 1 mm 1.59 mm
Each for $758.04
Only null left
 
06710393 DFS Item
View Documents
Restek™
RK19019
2 m 0.75 mm 0.95 mm
Each for $783.64
Only null left
 
06710392 DFS Item
View Documents
Restek™
RK19018
1 m 0.75 mm 0.95 mm
Each for $623.75
Only null left
 
Description

Description

  • Increased efficiency over traditional packed columns
  • Higher capacity than PLOT columns
  • Made from inert, flexible, SilcoSmooth tubing
  • Standard coils fit all instruments; no special instrument configurations required

Efficient, Inert, and Flexible

  • Unsurpassed inertness of SilcoSmooth tubing is based on Siltek deactivation
  • This allows the column to be flexed and coiled without any fear of chipping or cracking the inert surface

Easy to Install - Multiple Internal Diameters

  • Designed to fit packed and capillary injection systems
  • Standard wall (1/16 in. OD) micropacked columns offer improved efficiency in packed column instruments without the expense of converting to capillary injection systems
  • Smaller OD (0.95mm OD) micropacked columns install easily into a capillary injector, using slightly larger ferrules
  • Operate at flows exceeding 10 cc/min. for trouble-free operation

Braided Wire End Plugs

  • Braided wires are inserted into the column and secured by making a small crimp near the column outlet
  • End plugs are Siltek treated which makes the sample contacts only inert surfaces
  • Mesh Size: 100/120
  • Temperature Limits: Upto 275°C

Order Info

Custom packed and micropacked columns also available by request.

Specifications

Specifications

275°C
Stainless Steel
100/120
Hayesep Q
GC
HayeSep Q
S3
Videos
SDS
Documents

Documents